missing translation for 'onlineSavingsMsg'
Learn More

1-(4-chlorophenyl)-5-methyl-1H-pyrazole-4-carboxylic acid, 97%, Thermo Scientific™

Catalog Number p-2694342
Click to view available options
Quantity:
1 g
5 g
10 g
Packaging:
Amber glass bottle
This item is not returnable. View return policy
187998-35-2
C11H9ClN2O2
236.655
MFCD01566260
OAPQKGGVDCZENK-UHFFFAOYSA-N
2777166
1-(4-chlorophenyl)-5-methylpyrazole-4-carboxylic acid
CC1=C(C=NN1C2=CC=C(C=C2)Cl)C(=O)O
This item is not returnable. View return policy
187998-35-2
C11H9ClN2O2
236.655
MFCD01566260
OAPQKGGVDCZENK-UHFFFAOYSA-N
2777166
1-(4-chlorophenyl)-5-methylpyrazole-4-carboxylic acid
CC1=C(C=NN1C2=CC=C(C=C2)Cl)C(=O)O
CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber glass bottle
Quantity 1 g
Synonym 1-4-chlorophenyl-5-methyl-1h-pyrazole-4-carboxylic acid, 1-4-chlorophenyl-5-methylpyrazole-4-carboxylic acid, 1-4-chlorophenyl-5-methyl-1h-pyrazole-4-carboxylicacid, 4-carboxy-1-4-chlorophenyl-5-methyl-1h-pyrazole, 1h-pyrazole-4-carboxylicacid, 1-4-chlorophenyl-5-methyl, buttpark 9618-16, salor-int l317632-1ea, 1-4-chlorophenyl-5-methylpyrazole-4-carboxylic-acid, 4-chloro-4-carboxy-5-methyl-1h-pyrazol-1-yl benzene, 1h-pyrazole-4-carboxylicacid,1-4-chlorophenyl-5-methyl
Formula Weight 236.66
Percent Purity 97%
Chemical Name or Material 1-(4-Chlorophenyl)-5-methyl-1H-pyrazole-4-carboxylic acid
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'