missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 1-(4-Chlorophenyl)-3-[4-(trifluoromethyl)phenyl]urea, 97%

Brand:  Alfa Aesar™ H61537.03

Code : NEW

Additional Details : CAS Number : 2317-30-8 Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15416586

  • £59.78 / 1g


Estimated Shipment date
from Supplier 09-05-2018
Add to basket

Specifications

Chemical Identifiers

1-(4-Chlorophenyl)-3-[4-(trifluoromethyl)phenyl]urea
314.69
2668302
2317-30-8
MFCD00418718

Specifications

1-(4-Chlorophenyl)-3-[4-(trifluoromethyl)phenyl]urea
2317-30-8
97%
MFCD00418718
1g
Solid
314.69
2668302
Videos
SDS