missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 1-(4-Chlorophenyl)-3-[4-(trifluoromethoxy)phenyl]urea, 97%

Brand:  Alfa Aesar™ H64314.06

Code : NEW

Additional Details : CAS Number : 54730-71-1 Weight : 0.00500kg

Quantity 5g

 View more versions of this product

Product Code. 15478836

  • £331.78 / 5g


Estimated Shipment date
from Supplier 06-03-2018
Add to basket

Specifications

Chemical Identifiers

1-(4-Chlorophenyl)-3-[4-(trifluoromethoxy)phenyl]urea
330.69
2888769
54730-71-1
MFCD02632654

Specifications

1-(4-Chlorophenyl)-3-[4-(trifluoromethoxy)phenyl]urea
54730-71-1
97%
MFCD02632654
5g
Solid
330.69
2888769
Videos
SDS