missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 1-(4-Chlorophenyl)-3-(4-fluorophenyl)urea, 97%

Brand:  Alfa Aesar™ H62983.06

Code : NEW

Additional Details : CAS Number : 404-51-3 Weight : 0.00500kg

Quantity 5g

 View more versions of this product

Product Code. 15439418

  • £280.90 / 5g


Estimated Shipment date
from Supplier 31-01-2018
Add to basket

Specifications

Chemical Identifiers

1-(4-Chlorophenyl)-3-(4-fluorophenyl)urea
404-51-3
264.68
MFCD00018540

Specifications

1-(4-Chlorophenyl)-3-(4-fluorophenyl)urea
404-51-3
97%
MFCD00018540
5g
Solid
264.68
Videos
SDS