missing translation for 'onlineSavingsMsg'
Learn More

1-(4-Chlorophenyl)-1-cyclobutanecarboxylicacid 94%, ACROS Organics™

Brand:  Acros Organics 170720050

Code : 29

Additional Details : CAS Number : 50921-39-6 Weight : 0.05500kg


Quantity 5g
Packaging Glass bottle

 View more versions of this product

Product Code. 10732791

  • £12.30 / 5g


EU Stock 7
Add to basket


Specifications

Chemical Identifiers

1-(4-Chlorophenyl)-1-cyclobutanecarboxylic acid
94%
ClC6H4C4H6CO2H
94%
C11H11ClO2
210.66

Specifications

1-(4-Chlorophenyl)-1-cyclobutanecarboxylic acid
80.0° to 90.0°C
50921-39-6
Glass bottle
94%
C11H11ClO2
210.66
Light yellow to purple
5g
Authentic
Crystalline Powder
94%
ClC6H4C4H6CO2H
Videos
Safety and Handling

Safety and Handling

Product Identifier
  • 1-(4-Chlorophenyl)-1-cyclobutanecarboxylic acid

Signal Word
  • Warning

Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3

Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.

Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

SDS