missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1-(4-Chlorophenyl)-1-cyclobutanecarboxylic acid, 94%, Thermo Scientific™
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass Bottle
Specifications
Melting Point | 80.0°C to 90.0°C |
Color | Purple to Yellow |
Quantity | 100 g |
Infrared Spectrum | Authentic |
Packaging | Glass Bottle |
Physical Form | Crystalline Powder |
Assay Percent Range | 92% min. (HPLC) |
Linear Formula | ClC6H4C4H6CO2H |
Formula Weight | 210.66 |
Percent Purity | 94% |
Show More |
Safety and Handling
Product Identifier
- 1-(4-Chlorophenyl)-1-cyclobutanecarboxylic acid
- Warning
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
- H315-Causes skin irritation.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'