missing translation for 'onlineSavingsMsg'
Learn More

1-(4-Chlorobenzhydryl)piperazine, 90-95%, Thermo Scientific Chemicals

Catalog Number 10489780
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
This item is not returnable. View return policy
303-26-4
C17H19ClN2
286.79
UZKBSZSTDQSMDR-UHFFFAOYSA-N
1-4-chlorobenzhydryl piperazine, norchlorcyclizine, 1-4-chlorophenyl phenyl methyl piperazine, n-p-chlorobenzhydryl piperazine, 4-4-chlorobenzhydryl piperazine, 1-4-chlorophenyl phenylmethyl piperazine, n-4-chlorobenzhydryl piperazine, 1-4-chlorophenyl-phenylmethyl piperazine
9340
1-[(4-chlorophenyl)-phenylmethyl]piperazine
C1CN(CCN1)C(C2=CC=CC=C2)C3=CC=C(C=C3)Cl
This item is not returnable. View return policy
303-26-4
C17H19ClN2
286.79
UZKBSZSTDQSMDR-UHFFFAOYSA-N
1-4-chlorobenzhydryl piperazine, norchlorcyclizine, 1-4-chlorophenyl phenyl methyl piperazine, n-p-chlorobenzhydryl piperazine, 4-4-chlorobenzhydryl piperazine, 1-4-chlorophenyl phenylmethyl piperazine, n-4-chlorobenzhydryl piperazine, 1-4-chlorophenyl-phenylmethyl piperazine
9340
1-[(4-chlorophenyl)-phenylmethyl]piperazine
C1CN(CCN1)C(C2=CC=CC=C2)C3=CC=C(C=C3)Cl

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 65.0°C to 75.0°C
Color White to Beige
Boiling Point 178.0°C to 180.0°C (0.5 mmHg)
Flash Point >110°C
Infrared Spectrum Authentic
Assay Percent Range 90% min. (on dry substance) (Tertiary base)
Packaging Glass bottle
Quantity 25 g
Formula Weight 286.79
Percent Purity 90 to 95%
Physical Form Crystalline Powder
Chemical Name or Material 1-(4-Chlorobenzhydryl)piperazine
Show More Show Less
compliance-icons
Product Identifier
  • 1-(4-Chlorobenzhydryl)piperazine
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'