missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 1-[4-Chloro-3-(trifluoromethyl)phenyl]-3-(4-fluoro-2-methylphenyl)urea, 97%

Brand:  Alfa Aesar™ H65157.03

UNSPSC : 12352116

Code : NEW

Additional Details : Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15409426

  • £74.73 / 1g


Estimated Shipment date
from Supplier 06-07-2017
Add to basket

Specifications Description & Specifications

Chemical Identifiers

1-[4-Chloro-3-(trifluoromethyl)phenyl]-3-(4-fluoro-2-methylphenyl)urea
346.71
MFCD27975729

Specifications

1-[4-Chloro-3-(trifluoromethyl)phenyl]-3-(4-fluoro-2-methylphenyl)urea
Solid
346.71
1g
97%
MFCD27975729
Documentation

Technical Documents