missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Alfa Aesar™ 1-[4-Chloro-3-(trifluoromethyl)phenyl]-3-(4-ethylphenyl)urea, 97%
Brand: Alfa Aesar™ H65389.03
Code : NEW
Additional Details : Weight : 0.00100kg
Quantity | 1g |
---|
Chemical Identifiers
1-[4-Chloro-3-(trifluoromethyl)phenyl]-3-(4-ethylphenyl)urea | |
342.75 | |
MFCD07116106 |
Specifications
1-[4-Chloro-3-(trifluoromethyl)phenyl]-3-(4-ethylphenyl)urea | |
Solid | |
342.75 |
1g | |
97% | |
MFCD07116106 |