missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 1-[4-Chloro-3-(trifluoromethyl)phenyl]-3-[3-(trifluoromethyl)phenyl]urea, 97%

Brand:  Alfa Aesar™ H65760.03

Code : NEW

Additional Details : CAS Number : 370-51-4 Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15419886

  • £70.28 / 1g


Estimated Shipment date
from Supplier 02-07-2018
Add to basket

Specifications

Chemical Identifiers

1-[4-Chloro-3-(trifluoromethyl)phenyl]-3-[3-(trifluoromethyl)phenyl]urea
370-51-4
382.69
MFCD03416773

Specifications

1-[4-Chloro-3-(trifluoromethyl)phenyl]-3-[3-(trifluoromethyl)phenyl]urea
370-51-4
97%
MFCD03416773
1g
Solid
382.69
Videos
SDS