missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 1-[4-Chloro-3-(trifluoromethyl)phenyl]-3-(3-methoxyphenyl)urea, 97%

Brand:  Alfa Aesar™ H65660.03

Code : NEW

Additional Details : Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15459816

  • £79.71 / 1g


Estimated Shipment date
from Supplier 09-07-2018
Add to basket

Specifications

Chemical Identifiers

1-[4-Chloro-3-(trifluoromethyl)phenyl]-3-(3-methoxyphenyl)urea
344.72
MFCD06144443

Specifications

1-[4-Chloro-3-(trifluoromethyl)phenyl]-3-(3-methoxyphenyl)urea
Solid
344.72
1g
97%
MFCD06144443
Videos
SDS