missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 1-[4-Chloro-3-(trifluoromethyl)phenyl]-3-(3-isopropylphenyl)urea, 97%

Brand:  Alfa Aesar™ H65003.03

Code : NEW

Additional Details : Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15409316

  • £85.44 / 1g


Estimated Shipment date
from Supplier 12-03-2018
Add to basket

Specifications

Chemical Identifiers

1-[4-Chloro-3-(trifluoromethyl)phenyl]-3-(3-isopropylphenyl)urea
356.77
MFCD27975709

Specifications

1-[4-Chloro-3-(trifluoromethyl)phenyl]-3-(3-isopropylphenyl)urea
Solid
356.77
1g
97%
MFCD27975709
Videos
SDS