missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 1-[4-Chloro-3-(trifluoromethyl)phenyl]-3-(3,4-difluorophenyl)urea, 97%

Brand:  Alfa Aesar™ H65995.03

Code : NEW

Additional Details : Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15400087

  • £74.73 / 1g


Estimated Shipment date
from Supplier 05-06-2018
Add to basket

Specifications

Chemical Identifiers

1-[4-Chloro-3-(trifluoromethyl)phenyl]-3-(3,4-difluorophenyl)urea
350.67
MFCD27975852

Specifications

1-[4-Chloro-3-(trifluoromethyl)phenyl]-3-(3,4-difluorophenyl)urea
Solid
350.67
1g
97%
MFCD27975852
Videos
SDS