missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 1-[4-Chloro-3-(trifluoromethyl)phenyl]-3-(2-isopropylphenyl)urea, 97%

Brand:  Alfa Aesar™ H65944.03

Code : NEW

Additional Details : Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15470027

  • £79.71 / 1g


Estimated Shipment date
from Supplier 12-06-2018
Add to basket

Specifications

Chemical Identifiers

1-[4-Chloro-3-(trifluoromethyl)phenyl]-3-(2-isopropylphenyl)urea
356.77
MFCD27975843

Specifications

1-[4-Chloro-3-(trifluoromethyl)phenyl]-3-(2-isopropylphenyl)urea
Solid
356.77
1g
97%
MFCD27975843
Videos
SDS