missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 1-[4-Chloro-3-(trifluoromethyl)phenyl]-3-(2-fluorophenyl)urea, 97%

Brand:  Alfa Aesar™ H65673.03

Code : NEW

Additional Details : Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15459826

  • £56.92 / 1g


Estimated Shipment date
from Supplier 11-06-2018
Add to basket

Specifications

Chemical Identifiers

1-[4-Chloro-3-(trifluoromethyl)phenyl]-3-(2-fluorophenyl)urea
332.68
MFCD07060646

Specifications

1-[4-Chloro-3-(trifluoromethyl)phenyl]-3-(2-fluorophenyl)urea
Solid
332.68
1g
97%
MFCD07060646
Videos
SDS