missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 1-[4-Chloro-3-(trifluoromethyl)phenyl]-3-(2-fluorobenzyl)urea, 97%

Brand:  Alfa Aesar™ H65603.03

Code : NEW

Additional Details : Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15419776

  • £59.78 / 1g


Estimated Shipment date
from Supplier 07-06-2018
Add to basket

Specifications

Chemical Identifiers

1-[4-Chloro-3-(trifluoromethyl)phenyl]-3-(2-fluorobenzyl)urea
346.71
MFCD27975790

Specifications

1-[4-Chloro-3-(trifluoromethyl)phenyl]-3-(2-fluorobenzyl)urea
Solid
346.71
1g
97%
MFCD27975790
Videos
SDS