missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 1-[4-Chloro-3-(trifluoromethyl)phenyl]-3-(2,5-dichlorophenyl)urea, 97%

Brand:  Alfa Aesar™ H65260.03

UNSPSC : 12352116

Code : NEW

Additional Details : Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15409516

  • £91.90 / 1g


Estimated Shipment date
from Supplier 07-08-2017
Add to basket

Specifications Description & Specifications

Chemical Identifiers

1-[4-Chloro-3-(trifluoromethyl)phenyl]-3-(2,5-dichlorophenyl)urea
383.58
MFCD04289647

Specifications

1-[4-Chloro-3-(trifluoromethyl)phenyl]-3-(2,5-dichlorophenyl)urea
Solid
383.58
1g
97%
MFCD04289647
Documentation

Technical Documents