missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 1-[4-Chloro-3-(trifluoromethyl)phenyl]-3-(2,4-dimethylphenyl)urea, 97%

Brand:  Alfa Aesar™ H65923.03

Code : NEW

Additional Details : Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15430017

  • £74.73 / 1g


Estimated Shipment date
from Supplier 08-05-2018
Add to basket

Specifications

Chemical Identifiers

1-[4-Chloro-3-(trifluoromethyl)phenyl]-3-(2,4-dimethylphenyl)urea
342.75
MFCD06163178

Specifications

1-[4-Chloro-3-(trifluoromethyl)phenyl]-3-(2,4-dimethylphenyl)urea
Solid
342.75
1g
97%
MFCD06163178
Videos
SDS