missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 1-[4-Chloro-3-(trifluoromethyl)phenyl]-3-(2,3-dimethylphenyl)urea, 97%

Brand:  Alfa Aesar™ H65202.03

Code : NEW

Additional Details : Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15429456

  • £70.28 / 1g


Estimated Shipment date
from Supplier 06-02-2018
Add to basket

Specifications

Chemical Identifiers

1-[4-Chloro-3-(trifluoromethyl)phenyl]-3-(2,3-dimethylphenyl)urea
342.75
MFCD07062043

Specifications

1-[4-Chloro-3-(trifluoromethyl)phenyl]-3-(2,3-dimethylphenyl)urea
Solid
342.75
1g
97%
MFCD07062043
Videos
SDS