missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 1-(4-Chloro-2-nitrophenyl)pyrrolidine, 97%

Brand:  Alfa Aesar™ H63558.03

Code : NEW

Additional Details : CAS Number : 41173-36-8 Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15400319

  • £50.70 / 1g


Estimated Shipment date
from Supplier 01-02-2018
Add to basket

Specifications

Chemical Identifiers

1-(4-Chloro-2-nitrophenyl)pyrrolidine
41173-36-8
226.66
915044

Specifications

1-(4-Chloro-2-nitrophenyl)pyrrolidine
41173-36-8
97%
915044
1g
Solid
226.66
Videos
SDS