missing translation for 'onlineSavingsMsg'
Learn More

1-(4-Chloro-2-nitrophenyl)piperazine, 97%, Thermo Scientific™

Catalog Number 15440779
Click to view available options
Quantity:
250 mg
1 g
5 g
This item is not returnable. View return policy
405910-34-1
C10H12ClN3O2
241.675
MFCD00640777
NZFQOXSALNTRDH-UHFFFAOYSA-N
1-4-chloro-2-nitrophenyl piperazine, 4-chloro-2-nitrophenyl piperazine, 1-4-chloro-2-nitro-phenyl-piperazine, piperazine, 1-4-chloro-2-nitrophenyl
2735779
1-(4-chloro-2-nitrophenyl)piperazine
C1CN(CCN1)C2=C(C=C(C=C2)Cl)[N+](=O)[O-]
This item is not returnable. View return policy
405910-34-1
C10H12ClN3O2
241.675
MFCD00640777
NZFQOXSALNTRDH-UHFFFAOYSA-N
1-4-chloro-2-nitrophenyl piperazine, 4-chloro-2-nitrophenyl piperazine, 1-4-chloro-2-nitro-phenyl-piperazine, piperazine, 1-4-chloro-2-nitrophenyl
2735779
1-(4-chloro-2-nitrophenyl)piperazine
C1CN(CCN1)C2=C(C=C(C=C2)Cl)[N+](=O)[O-]

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Quantity 250 mg
Formula Weight 241.67
Percent Purity 97%
Physical Form Solid
Chemical Name or Material 1-(4-Chloro-2-nitrophenyl)piperazine
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'