missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 1-(4-Chloro-2-nitrophenyl)piperazine, 97%

Brand:  Alfa Aesar™ H63851.06

Code : NEW

Additional Details : CAS Number : 405910-34-1 Weight : 0.00500kg

Quantity 5g

 View more versions of this product

Product Code. 15430779

  • £202.50 / 5g


Estimated Shipment date
from Supplier 02-07-2018
Add to basket

Specifications

Chemical Identifiers

1-(4-Chloro-2-nitrophenyl)piperazine
405910-34-1
241.67
MFCD00640777

Spécification

1-(4-Chloro-2-nitrophenyl)piperazine
405910-34-1
97%
MFCD00640777
5g
Solid
241.67
Vidéos
FDS