missing translation for 'onlineSavingsMsg'
Learn More

1-(4-Chloro-2-nitrophenyl)-4-methylpiperazine, 97%, Thermo Scientific™

Catalog Number 15458486
Click to view available options
Quantity:
250 mg
1 g
5 g
This item is not returnable. View return policy
65639-68-1
C11H14ClN3O2
255.702
GYZYCENBDCHKGE-UHFFFAOYSA-N
1-4-chloro-2-nitrophenyl-4-methylpiperazine, piperazine, 1-4-chloro-2-nitrophenyl-4-methyl, cambridge id 5670746, 1-4-chloro-2-nitro-phenyl-4-methyl-piperazine
2860302
1-(4-chloro-2-nitrophenyl)-4-methylpiperazine
CN1CCN(CC1)C2=C(C=C(C=C2)Cl)[N+](=O)[O-]
This item is not returnable. View return policy
65639-68-1
C11H14ClN3O2
255.702
GYZYCENBDCHKGE-UHFFFAOYSA-N
1-4-chloro-2-nitrophenyl-4-methylpiperazine, piperazine, 1-4-chloro-2-nitrophenyl-4-methyl, cambridge id 5670746, 1-4-chloro-2-nitro-phenyl-4-methyl-piperazine
2860302
1-(4-chloro-2-nitrophenyl)-4-methylpiperazine
CN1CCN(CC1)C2=C(C=C(C=C2)Cl)[N+](=O)[O-]

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Quantity 250 mg
Formula Weight 255.7
Percent Purity 97%
Physical Form Solid
Chemical Name or Material 1-(4-Chloro-2-nitrophenyl)-4-methylpiperazine
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'