missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 1-(4-Chloro-2-methylphenyl)-3-[4-(trifluoromethoxy)phenyl]urea, 97%

Brand:  Alfa Aesar™ H64576.03

Code : NEW

Additional Details : Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15498986

  • £62.96 / 1g


Estimated Shipment date
from Supplier 05-02-2018
Add to basket

Specifications

Chemical Identifiers

1-(4-Chloro-2-methylphenyl)-3-[4-(trifluoromethoxy)phenyl]urea
344.72
MFCD26960705

Specifications

1-(4-Chloro-2-methylphenyl)-3-[4-(trifluoromethoxy)phenyl]urea
97%
344.72
Solid
1g
MFCD26960705
Videos
SDS