missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 1-(4-Chloro-2-methylphenyl)-3-(4-fluorophenyl)urea, 97%

Brand:  Alfa Aesar™ H62404.03

Code : NEW

Additional Details : Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15457396

  • £62.96 / 1g


Estimated Shipment date
from Supplier 12-06-2018
Add to basket

Specifications

Chemical Identifiers

1-(4-Chloro-2-methylphenyl)-3-(4-fluorophenyl)urea
278.71
MFCD02703560

Specifications

1-(4-Chloro-2-methylphenyl)-3-(4-fluorophenyl)urea
97%
278.71
Solid
1g
MFCD02703560
Videos
SDS