missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 1-(4-Chloro-2-methylphenyl)-3-(4-fluorophenyl)urea, 97%

Brand:  Alfa Aesar™ H62404.06

Code : NEW

Additional Details : Weight : 0.00500kg

Quantity 5g

 View more versions of this product

Product Code. 15467396

  • £314.82 / 5g


Estimated Shipment date
from Supplier 05-02-2018
Add to basket

Specifications

Chemical Identifiers

1-(4-Chloro-2-methylphenyl)-3-(4-fluorophenyl)urea
278.71
MFCD02703560

Specifications

1-(4-Chloro-2-methylphenyl)-3-(4-fluorophenyl)urea
97%
278.71
Solid
5g
MFCD02703560
Videos
SDS