missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 1-(4-Chloro-2-fluorophenyl)-3-[4-(trifluoromethyl)phenyl]urea, 97%

Brand:  Alfa Aesar™ H61966.03

Code : NEW

Additional Details : Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15406896

  • £85.44 / 1g


Estimated Shipment date
from Supplier 12-03-2018
Add to basket

Specifications

Chemical Identifiers

1-(4-Chloro-2-fluorophenyl)-3-[4-(trifluoromethyl)phenyl]urea
332.68
MFCD26960670

Specifications

1-(4-Chloro-2-fluorophenyl)-3-[4-(trifluoromethyl)phenyl]urea
Solid
332.68
1g
97%
MFCD26960670
Videos
SDS