missing translation for 'onlineSavingsMsg'
Learn More

1,4-Butanediphosphonic acid, 98%, ACROS Organics™

Brand:  Acros Organics 332200010

Code : 29

Additional Details : CAS Number : 4671-77-6 Weight : 0.05100kg Transport : UN number : 3261 Chem class : 8 Pack group : III


Packaging Glass bottle
Quantity 1g

 View more versions of this product

Product Code. 10051751

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Product Support team on 01509 555888 for assistance


Specifications

Chemical Identifiers

1, 4-Butanediphosphonic acid
97.5
98%
C4H12O6P2
218.08
4671-77-6
100.0
98%
(O)P(OH)2(CH2)4P(OH)2(O)
MFCD01631212

Specifications

1, 4-Butanediphosphonic acid
97.5
Authentic
Fine Powder
98%
(O)P(OH)2(CH2)4P(OH)2(O)
MFCD01631212
219.0° to 221.0°C
4671-77-6
100.0
Glass bottle
98%
C4H12O6P2
218.08
White
1g
Videos
Safety and Handling

Safety and Handling

Product Identifier
  • 1,4-Butanediphosphonic acid

Signal Word
  • Danger

Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B

Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.

Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

SDS