missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1,4-Butanediol diacrylate, 85+%, stab. with 50-105 ppm hydroquinone, Thermo Scientific Chemicals
Click to view available options
Quantity:
5 g
25 g
100 g
Description
1,4-Butanediol diacrylate is used as a multifunctional crosslinker. It is used in drying technology for paintings, coatings, inks, resins and adhesives. It serves as an intermediate product for polymer synthesis in the chemical industry.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Color | Colorless |
Density | 1.051 g/mL |
Boiling Point | 83°C (0.3 mmHg) |
Flash Point | >110°C (230°F) |
Odor | Pleasant |
Linear Formula | CH2=CHCO2(CH2)4O2CCH=CH2 |
Refractive Index | 1.456 |
Quantity | 5 g |
UN Number | UN1760 |
Beilstein | 1777487 |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'