missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 1-(4-Bromophenyl)cyclopropanecarboxylic acid, 98%

Brand:  Alfa Aesar™ H64163.MD

Code : NEW

Additional Details : CAS Number : 345965-52-8 Weight : 0.00025kg

Quantity 250mg

 View more versions of this product

Product Code. 15428746

  • £15.26 / 250mg


Estimated Shipment date
from Supplier 08-03-2018
Add to basket

Specifications

Chemical Identifiers

1-(4-Bromophenyl)cyclopropanecarboxylic acid
241.08
15699882
345965-52-8
MFCD07374439

Specifications

1-(4-Bromophenyl)cyclopropanecarboxylic acid
160° to 165°C
Solid
Off-white
MFCD07374439
345965-52-8
250mg
98%
241.08
15699882
Videos
SDS