missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1-(4-Bromophenoxy)hexane, 97%, Thermo Scientific™
Click to view available options
Quantity:
1 g
10 g
50 g
Specifications
Assay Percent Range | 97% |
Quantity | 1 g |
Formula Weight | 257.17 |
Chemical Name or Material | 1-(4-Bromophenoxy)hexane |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'