Learn More
1-[4-(Bromomethyl)phenyl]-1H-pyrazole, 95%, Maybridge
Brand: Maybridge CC18408DE
Code : 31
Additional Details : CAS Number : 368869-85-6 Weight : 0.05500kg Transport : UN number : 1759 Chem class : 1a Pack group : III
Quantity | 5g |
---|---|
Packaging | Amber Glass Bottle |
Chemical Identifiers
1-[4-(Bromomethyl)phenyl]-1H-pyrazole | |
95% | |
237.1 |
95% | |
C10H9BrN2 |
Specifications
1-[4-(Bromomethyl)phenyl]-1H-pyrazole | |
Amber Glass Bottle | |
95% | |
C10H9BrN2 |
368869-85-6 | |
5g | |
95% | |
237.1 |