missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Alfa Aesar™ 1-(4-Bromobenzyl)-3-[4-chloro-3-(trifluoromethyl)phenyl]urea, 97%
Brand: Alfa Aesar™ H65493.03
Code : NEW
Additional Details : Weight : 0.00100kg
Quantity | 1g |
---|
Chemical Identifiers
1-(4-Bromobenzyl)-3-[4-chloro-3-(trifluoromethyl)phenyl]urea | |
407.62 | |
MFCD27975768 |
Specifications
1-(4-Bromobenzyl)-3-[4-chloro-3-(trifluoromethyl)phenyl]urea | |
Solid | |
407.62 |
1g | |
97% | |
MFCD27975768 |