missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 1-(4-Bromo-3-methylphenyl)-3-[4-(trifluoromethoxy)phenyl]urea, 97%

Brand:  Alfa Aesar™ H64681.06

Code : NEW

Additional Details : Weight : 0.00500kg

Quantity 5g

 View more versions of this product

Product Code. 15499066

  • £221.54 / 5g


Estimated Shipment date
from Supplier 08-06-2018
Add to basket

Specifications

Chemical Identifiers

1-(4-Bromo-3-methylphenyl)-3-[4-(trifluoromethoxy)phenyl]urea
389.17
MFCD26960708

Specifications

1-(4-Bromo-3-methylphenyl)-3-[4-(trifluoromethoxy)phenyl]urea
5g
389.17
Solid
97%
MFCD26960708
Videos
SDS