missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 1-(4-Bromo-2-methylphenyl)-3-[4-(trifluoromethoxy)phenyl]urea, 97%

Brand:  Alfa Aesar™ H64455.03

Code : NEW

Additional Details : Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15418936

  • £46.00 / 1g


Estimated Shipment date
from Supplier 12-06-2018
Add to basket

Specifications

Chemical Identifiers

1-(4-Bromo-2-methylphenyl)-3-[4-(trifluoromethoxy)phenyl]urea
389.17
MFCD26960702

Specifications

1-(4-Bromo-2-methylphenyl)-3-[4-(trifluoromethoxy)phenyl]urea
1g
389.17
Solid
97%
MFCD26960702
Videos
SDS