missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Alfa Aesar™ 1-(4-Bromo-2-methylphenyl)-3-(4-fluorophenyl)urea, 97%
Brand: Alfa Aesar™ H62418.06
Code : NEW
Additional Details : Weight : 0.00500kg
Quantity | 5g |
---|
Chemical Identifiers
1-(4-Bromo-2-methylphenyl)-3-(4-fluorophenyl)urea | |
323.16 | |
MFCD07072623 |
Specifications
1-(4-Bromo-2-methylphenyl)-3-(4-fluorophenyl)urea | |
5g | |
323.16 |
Solid | |
97% | |
MFCD07072623 |