missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 1-(4-Bromo-2-fluorophenyl)-3-[4-(trifluoromethyl)phenyl]urea, 97%

Brand:  Alfa Aesar™ H61935.03

Code : NEW

Additional Details : Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15486866

  • £95.61 / 1g


Estimated Shipment date
from Supplier 11-06-2018
Add to basket

Specifications

Chemical Identifiers

1-(4-Bromo-2-fluorophenyl)-3-[4-(trifluoromethyl)phenyl]urea
377.13
MFCD24849001

Specifications

1-(4-Bromo-2-fluorophenyl)-3-[4-(trifluoromethyl)phenyl]urea
Solid
1g
MFCD24849001
White
97%
377.13
Videos
SDS