missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 1-(4-Bromo-2-fluorophenyl)-3-(4-fluorophenyl)urea, 97%

Brand:  Alfa Aesar™ H62101.03

Code : NEW

Additional Details : Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15427026

  • £54.38 / 1g


Estimated Shipment date
from Supplier 10-05-2018
Add to basket

Specifications

Chemical Identifiers

1-(4-Bromo-2-fluorophenyl)-3-(4-fluorophenyl)urea
327.12
MFCD07060296

Specifications

1-(4-Bromo-2-fluorophenyl)-3-(4-fluorophenyl)urea
97%
327.12
Solid
1g
MFCD07060296
Videos
SDS