missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 1-(4-Bromo-2-chlorophenyl)-3-(4-fluorophenyl)urea, 97%

Brand:  Alfa Aesar™ H62714.03

Code : NEW

Additional Details : Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15447766

  • £56.29 / 1g


Estimated Shipment date
from Supplier 09-07-2018
Add to basket

Specifications

Chemical Identifiers

1-(4-Bromo-2-chlorophenyl)-3-(4-fluorophenyl)urea
343.58
MFCD07029641

Specifications

1-(4-Bromo-2-chlorophenyl)-3-(4-fluorophenyl)urea
97%
MFCD07029641
Solid
343.58
1g
Videos
SDS