missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1,4-Bis(trimethylsilyl)-1,3-butadiyne, 98%, Thermo Scientific Chemicals
49.20 GBP valid until 2024-03-29
Use promo code "21615" to get your promotional price.
Use promo code "21615" to get your promotional price.
Click to view available options
Quantity:
1 g
5 g
Description
1,4-Bis(trimethylsilyl)-1,3-butadiyne is used in Negishi protocol for the synthesis of glycosylated oligo(ethynylene)s. It is also used as pharmaceutical intermediates.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
1,4-Bis(trimethylsilyl)-1,3-butadiyne is used in Negishi protocol for the synthesis of glycosylated oligo(ethynylene)s. It is also used as pharmaceutical intermediates.
Solubility
Insoluble in water.
Notes
Store at 4°C. Incompatible materials are Strong oxidizing agents. Store in cool place. Keep container tightly closed in a dry and well-ventilated place.
1,4-Bis(trimethylsilyl)-1,3-butadiyne is used in Negishi protocol for the synthesis of glycosylated oligo(ethynylene)s. It is also used as pharmaceutical intermediates.
Solubility
Insoluble in water.
Notes
Store at 4°C. Incompatible materials are Strong oxidizing agents. Store in cool place. Keep container tightly closed in a dry and well-ventilated place.
Specifications
Melting Point | 110°C to 113°C |
Quantity | 5 g |
UN Number | UN1325 |
Beilstein | 1758268 |
Synonym | 1,4-bis trimethylsilyl-1,3-butadiyne, 1,4-bis trimethylsilyl butadiyne, 1,4-bis trimethylsilyl buta-1,3-diyne, trimethyl 4-trimethylsilylbuta-1,3-diynyl silane, btmsbd, acmc-1ctvu, bis trimethylsiyl butadiyne, ksc491i6r, bis trimethylsilyl-1,3-butadiyne, bis trimethylsilyl butadiyne |
Solubility Information | Insoluble in water. |
IUPAC Name | trimethyl(4-trimethylsilylbuta-1,3-diynyl)silane |
PubChem CID | 138279 |
Formula Weight | 194.43 |
Percent Purity | 98% |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'