missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Alfa Aesar™ 1,4-Bis[alpha-(4-chlorophenyl)benzyl]piperazine dihydrochloride, 97%
Brand: Alfa Aesar™ H63961.06
Code : NEW
Additional Details : Weight : 0.00500kg
Quantity | 5g |
---|
Chemical Identifiers
1,4-Bis[alpha-(4-chlorophenyl)benzyl]piperazine dihydrochloride | |
560.38 | |
1,4-Bis[(4-chlorophenyl)phenylmethyl]piperazine dihydrochloride |
346451-15-8 | |
MFCD03844641 |
Specifications
1,4-Bis[alpha-(4-chlorophenyl)benzyl]piperazine dihydrochloride | |
Solid | |
560.38 | |
White | |
1,4-Bis[(4-chlorophenyl)phenylmethyl]piperazine dihydrochloride |
346451-15-8 | |
97% | |
MFCD03844641 | |
5g |