missing translation for 'onlineSavingsMsg'
Learn More

1,4-Bis[alpha-(4-chlorophenyl)benzyl]piperazine dihydrochloride, 97%, Thermo Scientific Chemicals

Catalog Number p-7055393
Click to view available options
Quantity:
250 mg
1 g
5 g
This item is not returnable. View return policy
346451-15-8
C30H30Cl4N2
560.384
MFCD03844641
DLEQNCKNBPYHEY-UHFFFAOYSA-N
1,4-bis 4-chlorophenyl phenylmethyl piperazine dihydrochloride, cetirizine impurity d, 1,4-bis 4-chlorophenyl phenylmethyl-piperazine dihydrochloride, 1,4-bis 4-chlorophenyl phenyl methyl piperazine dihydrochloride, 1,4-bis 4-chlorophenyl-phenylmethyl piperazine;dihydrochloride
24720891
1,4-bis[(4-chlorophenyl)-phenylmethyl]piperazine;dihydrochloride
C1CN(CCN1C(C2=CC=CC=C2)C3=CC=C(C=C3)Cl)C(C4=CC=CC=C4)C5=CC=C(C=C5)Cl.Cl.Cl
This item is not returnable. View return policy
346451-15-8
C30H30Cl4N2
560.384
MFCD03844641
DLEQNCKNBPYHEY-UHFFFAOYSA-N
1,4-bis 4-chlorophenyl phenylmethyl piperazine dihydrochloride, cetirizine impurity d, 1,4-bis 4-chlorophenyl phenylmethyl-piperazine dihydrochloride, 1,4-bis 4-chlorophenyl phenyl methyl piperazine dihydrochloride, 1,4-bis 4-chlorophenyl-phenylmethyl piperazine;dihydrochloride
24720891
1,4-bis[(4-chlorophenyl)-phenylmethyl]piperazine;dihydrochloride
C1CN(CCN1C(C2=CC=CC=C2)C3=CC=C(C=C3)Cl)C(C4=CC=CC=C4)C5=CC=C(C=C5)Cl.Cl.Cl

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Color White
Quantity 250 mg
Formula Weight 560.38
Percent Purity 97%
Physical Form Solid
Chemical Name or Material 1,4-Bis[alpha-(4-chlorophenyl)benzyl]piperazine dihydrochloride

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'