missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 1,4-Bis(5-phenyloxazol-2-yl)benzene, 98+%

Brand:  Alfa Aesar™ A12321.06

Code : NEW

Additional Details : CAS Number : 1806-34-4 Weight : 0.00500kg

Quantity 5g

 View more versions of this product

Product Code. 11429433

  • £27.90 / 5g


EU Stock 1
Add to basket

Specifications

Chemical Identifiers

1,4-Bis (5-phenyloxazol-2-yl)benzene
C24H16N2O2
MFCD00005309
POPOP
1806-34-4
364.41
325741

Specifications

1,4-Bis (5-phenyloxazol-2-yl)benzene
≥98%
364.41
325741
241° to 243°C
1806-34-4
C24H16N2O2
MFCD00005309
POPOP
5g
Videos
SDS