missing translation for 'onlineSavingsMsg'
Learn More

1,4-Benzodioxane, 98%, Thermo Scientific Chemicals

Catalog Number p-7023722
Click to view available options
Quantity:
10 g
50 g
250 g
This item is not returnable. View return policy
493-09-4
C8H8O2
136.15
MFCD00006821
BNBQRQQYDMDJAH-UHFFFAOYSA-N
1,4-benzodioxane, 1,4-benzodioxan, 2,3-dihydrobenzo b 1,4 dioxine, 1,4-benzodioxin, 2,3-dihydro, pyrocatechol ethylene ether, 1,2-ethylenedioxy benzene, benzodioxan, ethylene o-phenylene dioxide, 1,2-ethylenedioxybenzene, 1, 4-benzdioxan
10301
2,3-dihydro-1,4-benzodioxine
C1COC2=CC=CC=C2O1
This item is not returnable. View return policy
493-09-4
C8H8O2
136.15
MFCD00006821
BNBQRQQYDMDJAH-UHFFFAOYSA-N
1,4-benzodioxane, 1,4-benzodioxan, 2,3-dihydrobenzo b 1,4 dioxine, 1,4-benzodioxin, 2,3-dihydro, pyrocatechol ethylene ether, 1,2-ethylenedioxy benzene, benzodioxan, ethylene o-phenylene dioxide, 1,2-ethylenedioxybenzene, 1, 4-benzdioxan
10301
2,3-dihydro-1,4-benzodioxine
C1COC2=CC=CC=C2O1

Benzo-1,4-dioxane (2,3-dihydro-1,4-benzodioxin) is used in the synthesis of stereoisomers which were evaluated as α- and β- adrenergic antagonists

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Benzo-1,4-dioxane (2,3-dihydro-1,4-benzodioxin) is used in the synthesis of stereoisomers which were evaluated as α- and β- adrenergic antagonists

Solubility
Slightly soluble in water.

Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
TRUSTED_SUSTAINABILITY
Density 1.17
Boiling Point 216°C
Flash Point 87°C (188°F)
Refractive Index 1.549
Quantity 10 g
Beilstein 120846
Solubility Information Slightly soluble in water.
Formula Weight 136.15
Percent Purity 98%
Chemical Name or Material 1,4-Benzodioxane

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'