missing translation for 'onlineSavingsMsg'
Learn More

1,4-Benzodioxan-5-carboxylic acid, 98%, ACROS Organics™

Brand:  Acros Organics 399610050

Code : 29

Additional Details : CAS Number : 4442-53-9 Weight : 0.05500kg


Quantity 5g

 View more versions of this product

Product Code. 10504944

  • £89.40 / 5g


Estimated Shipment
20-06-2018
Add to basket


Specifications

Chemical Identifiers

1, 4-Benzodioxan-5-carboxylic acid
98%
MFCD00239415
4442-53-9
C9H8O4

Specifications

1, 4-Benzodioxan-5-carboxylic acid
98%
MFCD00239415
4442-53-9
C9H8O4
5g
Videos
Safety and Handling

Safety and Handling

Product Identifier
  • 1,4-Benzodioxan-5-carboxylic acid

Signal Word
  • Warning

Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3

Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.

Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

SDS