Learn More
Thermo Scientific Maybridge 1,3-Thiazole-2-carbonyl chloride, 1g, 90%
Brand: Thermo Scientific Maybridge CC09502DA
1,3-Thiazole-2-carbonyl chloride, 1g, 90%
Code : 31
Additional Details : CAS Number : 30216-57-0 Weight : 0.05100kg Transport : UN number : 3261 Chem class : 8 Pack group : II