missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1,3-Thiazol-2-ylmethanol, 97+%, Thermo Scientific™
Click to view available options
Quantity:
1 g
5 g
10 g
Packaging:
Amber Glass Bottle
Specifications
Assay Percent Range | 97% |
Packaging | Amber Glass Bottle |
Quantity | 1 g |
IUPAC Name | 1,3-thiazol-2-ylmethanol |
Formula Weight | 115.16 |
Percent Purity | 97% |
Chemical Name or Material | 1, 3-Thiazol-2-ylmethanol |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'