missing translation for 'onlineSavingsMsg'
Learn More

1-(3-Pyridylmethyl)-1H-pyrazole-4-boronic acid pinacol ester, Thermo Scientific™

Catalog Number p-7031023
Click to view available options
Quantity:
1 g
This item is not returnable. View return policy
864754-21-2
C15H20BN3O2
285.15
MFCD08706029
LVLQAPQDEZOFQU-UHFFFAOYSA-N
3-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-1h-pyrazol-1-yl methyl pyridine, 1-pyridin-3-ylmethyl-1h-pyrazole-4-boronic acid, pinacol ester, 3-4-4,4,5,5-tetramethyl-1,3,2 dioxaborolan-2-yl-pyrazol-1-ylmethyl-pyridine, 1-3-pyridylmethyl-1h-pyrazole-4-boronic acid pinacol ester, 3-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl pyrazol-1-yl methyl pyridine, 3-4-tetramethyl-1,3,2-dioxaborolan-2-yl pyrazol-1-yl methyl pyridine, pubchem18546, 1-pyridin-3-ylmethyl-1h-pyrazole-4-boronic acid
17999185
3-[[4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyrazol-1-yl]methyl]pyridine
CC1(C)OB(OC1(C)C)C1=CN(CC2=CN=CC=C2)N=C1
This item is not returnable. View return policy
864754-21-2
C15H20BN3O2
285.15
MFCD08706029
LVLQAPQDEZOFQU-UHFFFAOYSA-N
3-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-1h-pyrazol-1-yl methyl pyridine, 1-pyridin-3-ylmethyl-1h-pyrazole-4-boronic acid, pinacol ester, 3-4-4,4,5,5-tetramethyl-1,3,2 dioxaborolan-2-yl-pyrazol-1-ylmethyl-pyridine, 1-3-pyridylmethyl-1h-pyrazole-4-boronic acid pinacol ester, 3-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl pyrazol-1-yl methyl pyridine, 3-4-tetramethyl-1,3,2-dioxaborolan-2-yl pyrazol-1-yl methyl pyridine, pubchem18546, 1-pyridin-3-ylmethyl-1h-pyrazole-4-boronic acid
17999185
3-[[4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyrazol-1-yl]methyl]pyridine
CC1(C)OB(OC1(C)C)C1=CN(CC2=CN=CC=C2)N=C1

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Melting Point 102°C to 106°C
Quantity 1 g
Formula Weight 285.16
Chemical Name or Material 1-(3-Pyridylmethyl)-1H-pyrazole-4-boronic acid pinacol ester
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'