missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1-(3-Methylpyrazin-2-yl)ethan-1-one, 97%, Thermo Scientific™
Click to view available options
Quantity:
10 g
25 g
Specifications
CAS Min % | 97.0 |
Density | 1.1100g/mL |
Assay Percent Range | 97% |
Packaging | Amber Glass Bottle |
Quantity | 10 g |
Formula Weight | 136.15 |
Percent Purity | 97% |
Chemical Name or Material | 1-(3-Methylpyrazin-2-yl)ethan-1-one |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'