Learn More
1-(3-Methylphenyl)piperazine, 99%, ACROS Organics™
Brand: Acros Organics 345950050
Code : 50
Additional Details : CAS Number : 41186-03-2 Weight : 0.06000kg Transport : UN number : 2735 Chem class : 8 Pack group : III
Packaging | Glass bottle |
---|---|
Quantity | 5g |
Chemical Identifiers
41186-03-2 | |
176.26 | |
JIWHIRLNKIUYSM-UHFFFAOYSA-N | |
83111 | |
CC1=CC(=CC=C1)N2CCNCC2 |
C11H16N2 | |
MFCD00040734 | |
1-3-methylphenyl piperazine, 1-m-tolyl piperazine, 1-m-tolylpiperazine, 1-3-methylphenyl-piperazine, 1-m-tolyl-piperazine, piperazine, 1-3-methylphenyl, 3-methylphenyl piperazine, 4-m-tolylpiperazine, 1-m-tolyl-piperzine, 4-m-tolyl-piperazine | |
1-(3-methylphenyl)piperazine |
Specifications
1-(3-Methylphenyl)piperazine | |
Glass bottle | |
Colorless to Yellow | |
99% | |
98.5 | |
98.5% min. (GC) | |
MFCD00040734 | |
JIWHIRLNKIUYSM-UHFFFAOYSA-N | |
1-(3-methylphenyl)piperazine | |
83111 | |
Viscous Liquid |
Authentic | |
1.5755 to 1.5785 | |
5g | |
41186-03-2 | |
100.0 | |
C11H16N2 | |
1-3-methylphenyl piperazine, 1-m-tolyl piperazine, 1-m-tolylpiperazine, 1-3-methylphenyl-piperazine, 1-m-tolyl-piperazine, piperazine, 1-3-methylphenyl, 3-methylphenyl piperazine, 4-m-tolylpiperazine, 1-m-tolyl-piperzine, 4-m-tolyl-piperazine | |
CC1=CC(=CC=C1)N2CCNCC2 | |
176.26 | |
176.26 | |
99% |
Safety and Handling
- 1-(3-Methylphenyl)piperazine
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 1
- Skin corrosion/irritation Category 1 C
Hazard Statement
- H318-Causes serious eye damage.
- H314-Causes severe skin burns and eye damage.
- H312-Harmful in contact with skin.
- H302-Harmful if swallowed.
Precautionary Statement
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P264-Wash thoroughly after handling.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.